DTXSID30746626
Catalog #: I-1595.0050BA
Supplier: Bachem Americas
CAS Number: 133448-21-2
Specifications
- Size:50 mg
- Cat. no.:I-1595.0050BA
- Supplier No.:I-1595.0050
Specifications
About this item
Synonyms: Boc-Gln-Gly-Arg-AMC·HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 653.134 g/mol
Formula:C₂₈H₄₁ClN₈O₈
MW:653.134 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00133632
CAS Number:133448-21-2
Customers Who Bought This Also Bought