Boc-Leu-Lys-Arg-MCA
Catalog #: I-1570.0050BA
Supplier: Bachem Americas
CAS Number: 109358-47-6
Specifications
- Size:50 mg
- Cat. no.:I-1570.0050BA
- Supplier No.:I-1570.0050
Specifications
About this item
Synonyms: Boc-Leu-Lys-Arg-AMC
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 672.828 g/mol
Formula:C₃₃H₅₂N₈O₇
MW:672.828 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00133640
CAS Number:109358-47-6
Customers Who Bought This Also Bought