Boc-Gly-Arg-Arg-AMC
Supplier: Bachem Americas
Certificates
About this item
Formula:C₂₉H₄₄N₁₀O₇
MDL Number:MFCD02259611
CAS Number:113866-14-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 644.734 g/mol
:Boc-Gly-Arg-Arg-AMC
Storage Temperature:Freezer
Customers Who Bought This Also Bought