Boc-Gln-Ala-Arg-AMC · HCl
Supplier: Bachem Americas
Certificates
Certificates
About this item
Boc-QAR-AMC, highly reactive substrate for trypsin. Fluorogenic substrate for prostasin and matriptase.
Customers Who Bought This Also Bought
Water, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
334,23 $ to 2 005,34 $
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
314,91 $
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
1 313,27 $ to 5 149,78 $
ART® 100E Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
441,38 $ to 1 728,95 $
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
464,49 $