Boc-Glu(obzl)-Gly-Arg-Mca hydrochloride
Catalog #: I-1545.0050BA
Supplier: Bachem Americas
CAS Number: 133448-22-3
Specifications
- Size:50 mg
- Cat. no.:I-1545.0050BA
- Supplier No.:I-1545.0050
Specifications
About this item
Synonyms: Boc-Glu(OBzl)-Gly-Arg-AMC·HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 744.243 g/mol
Formula:C₃₅H₄₆ClN₇O₉
MW:744.243 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00133631
CAS Number:133448-22-3
Customers Who Bought This Also Bought