A-4501
Catalog #: I-1030.0250BA
Supplier: Bachem Americas
CAS Number: 201847-54-3
Specifications
- Size:250 mg
- Cat. no.:I-1030.0250BA
- Supplier No.:I-1030.0250
Specifications
About this item
Synonyms: H-β-Ala-AMC·TFA
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 360.289 g/mol
Formula:C₁₅H₁₅F₃N₂O₅
MW:360.289 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00151970
CAS Number:201847-54-3
Customers Who Bought This Also Bought