Ac-Lys-AMC (acetate salt)
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Ac-Lys-AMC
Formula:C₁₈H₂₃N₃O₄
MW:345.399 g/mol
MDL Number:MFCD00151959
CAS Number:156661-42-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 345.399 g/mol
:Ac-Lys-AMC (acetate salt)
Storage Temperature:Freezer
Customers Who Bought This Also Bought