Ac-D-Lys-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-alpha-acetyl-D-lysine
Formula:C₈H₁₆N₂O₃
MW:188.227 g/mol
MDL Number:MFCD00066056
CAS Number:58840-79-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 188.227 g/mol
Storage Temperature:Ambient
Customers Who Bought This Also Bought