Ac-D-Arg-OH
Catalog #: F-4085.0005BA
Supplier: Bachem Americas
CAS Number: 2389-86-8
Specifications
- Size:5 g
- Cat. no.:F-4085.0005BA
- Supplier No.:F-4085.0005
Specifications
About this item
:Ac-D-Arg-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 216,24 g/mol
Formula:C₈H₁₆N₄O₃
Storage Temperature:Ambient
MDL Number:MFCD00272231
CAS Number:2389-86-8
EINECS:219-225-2
Customers Who Bought This Also Bought