3-(4-nitrophenyl)-L-alanine ethyl ester hydrochloride
Catalog #: F-4080.0025BA
Supplier: Bachem Americas
CAS Number: 58816-66-3
Specifications
- Size:25 g
- Cat. no.:F-4080.0025BA
- Supplier No.:F-4080.0025
Specifications
About this item
Synonyms: H-Phe(4-NO2)-OEt·HCl,H-Phe(4-NO2)-OEt.HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 274.701 g/mol
Formula:C₁₁H₁₄N₂O₄·HCl
MW:274.701 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD08275409
CAS Number:58816-66-3
EINECS:261-455-0
Customers Who Bought This Also Bought