2-Acetamidosuccinic acid
Catalog #: F-4070.0005BA
Supplier: Bachem Americas
CAS Number: 2545-40-6
Specifications
- Size:5 g
- Cat. no.:F-4070.0005BA
- Supplier No.:F-4070.0005
Specifications
About this item
Synonyms: Ac-DL-Asp-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 175.14 g/mol
Formula:C₆H₉NO₅
MW:175.14 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00063183
CAS Number:2545-40-6
EINECS:219-827-5
Customers Who Bought This Also Bought