Fmoc-4-(neopentyloxysulfonyl)-ABU-OH
Catalog #: F-4040.0001BA
Supplier: Bachem Americas
CAS Number: 220951-81-5
Specifications
- Size:1 g
- Cat. no.:F-4040.0001BA
- Supplier No.:F-4040.0001
Specifications
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 475,556 g/mol
Formula:C₂₄H₂₉NO₇S
Storage Temperature:Refrigerator
MDL Number:MFCD03424225
CAS Number:220951-81-5
Customers Who Bought This Also Bought