Ethyl-4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride
Catalog #: F-3480.0001BA
Supplier: Bachem Americas
CAS Number: 180258-46-2
Specifications
- Size:1 g
- Cat. no.:F-3480.0001BA
- Supplier No.:F-3480.0001
Specifications
About this item
Synonyms: Ethyl 4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 205.642 g/mol
Formula:C₇H₁₂ClN₃O₂
MW:205.642 g/mol
Storage Temperature:Freezer
MDL Number:MFCD01318760
CAS Number:180258-46-2
Customers Who Bought This Also Bought