(2S,4R)-2-amino-4-hydroxypentanedioic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: H-(2S
Formula:C₅H₉NO₅
MW:163.129 g/mol
MDL Number:MFCD00672375
CAS Number:2485-33-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 163.129 g/mol
:(2S,4R)-2-amino-4-hydroxypentanedioic acid
Storage Temperature:Freezer
Customers Who Bought This Also Bought