D-Valine methyl ester hydrochloride
Catalog #: F-3160.0005BA
Supplier: Bachem Americas
CAS Number: 7146-15-8
Specifications
- Size:5 g
- Cat. no.:F-3160.0005BA
- Supplier No.:F-3160.0005
Specifications
About this item
Synonyms: H-D-Val-OMe.HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 167.633 g/mol
Formula:C₆H₁₃NO₂
MW:167.633 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD00237309
CAS Number:21685-47-2
Customers Who Bought This Also Bought