(H-Homocys-OH)₂
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (2S)-2-Amino-4-[[(3S)-3-amino-4-hydroxy-4-oxo-butyl]disulphanyl]butanoic acid,(H-HoCys-OH)2
Formula:C₈H₁₆N₂O₄S₂
MW:268.36 g/mol
MDL Number:MFCD00020391
CAS Number:626-72-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 268.36 g/mol
Storage Temperature:Ambient
EINECS:210-962-5
Customers Who Bought This Also Bought