H-4,5-Dehydro-Leu-OH
Catalog #: F-2985.0001BA
Supplier: Bachem Americas
CAS Number: 87392-13-0
Specifications
- Size:1 g
- Cat. no.:F-2985.0001BA
- Supplier No.:F-2985.0001
Specifications
About this item
Synonyms: (S)-2-Methallylglycine
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 129.159 g/mol
:H-4,5-Dehydro-Leu-OH
Formula:C₆H₁₁NO₂
MW:129.159 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00237695
CAS Number:87392-13-0
Customers Who Bought This Also Bought