Methyl-2-amino-2-methyl-3-phenylpropanoate hydrochloride
Catalog #: F-2805.0005BA
Supplier: Bachem Americas
CAS Number: 64665-60-7
Specifications
- Size:5 g
- Cat. no.:F-2805.0005BA
- Supplier No.:F-2805.0005
Specifications
About this item
Synonyms: Methyl 2-amino-2-methyl-3-phenylpropanoate hydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 229.704 g/mol
:Methyl-2-amino-2-methyl-3-phenylpropanoate hydrochloride
Formula:C₁₁H₁₆ClNO₂
MW:229.704 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD00237767
CAS Number:64665-60-7
Customers Who Bought This Also Bought