Eflornithine hydrochloride monohydrate
Supplier: Bachem Americas
Certificates
About this item
Synonyms: DL-alpha-Difluoromethylornithine Hydrochloride,2
Formula:C₆H₁₅ClF₂N₂O₃
MW:236,64 g/mol
MDL Number:MFCD00242734
CAS Number:96020-91-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 236,64 g/mol
:Eflornithine hydrochloride monohydrate
Storage Temperature:Freezer
Specification Test Results
- Eflornithine hydrochloride monohydrate:
Specification Test Results
Customers Who Bought This Also Bought