4-Nitro-DL-phenylalanine
Catalog #: F-1905.0005BA
Supplier: Bachem Americas
CAS Number: 2922-40-9
Specifications
- Size:5 g
- Cat. no.:F-1905.0005BA
- Supplier No.:F-1905.0005
Specifications
Specification Test Results
- 4-Nitro-DL-phenylalanine:
Specification Test Results
About this item
Synonyms: H-p-Nitro-DL-Phe-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 210.189 g/mol
Formula:C₉H₁₀N₂O₄
MW:210.189 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00007384
CAS Number:2922-40-9
EINECS:220-868-6
Customers Who Bought This Also Bought