β-Chloro-D-alanine hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (S)-2-Amino-3-chloropropanoic acid hydrochloride
Formula:C₃H₇Cl₂NO₂
MW:159.994 g/mol
MDL Number:MFCD00012616
CAS Number:51887-88-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 159.994 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought