2-Amino-4,4-dimethylpentanoic acid
Catalog #: F-1325.0005BA
Supplier: Bachem Americas
CAS Number: 106247-35-2
Specifications
- Size:5 g
- Cat. no.:F-1325.0005BA
- Supplier No.:F-1325.0005
Specifications
About this item
Synonyms: H-β-tBu-DL-Ala-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 145.202 g/mol
:2-Amino-4,4-dimethylpentanoic acid
Formula:C₇H₁₅NO₂
MW:145.202 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00066078
CAS Number:106247-35-2
Customers Who Bought This Also Bought