γ-Methyl-L-leucine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: H-β-tBu-Ala-OH,(S)-2-Amino-4
Formula:C₇H₁₅NO₂
MW:145.202 g/mol
MDL Number:MFCD00066079
CAS Number:57224-50-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 145.202 g/mol
Storage Temperature:Ambient
Customers Who Bought This Also Bought