4-Bromo-DL-phenylalanine
Supplier: Bachem Americas
Certificates
About this item
Formula:C₉H₁₀BrNO₂
MW:244.09
MDL Number:MFCD00002599
CAS Number:14091-15-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 244.09
Storage Temperature:Ambient
EINECS:237-941-3
Customers Who Bought This Also Bought