O-Benzyl-D-Serine
Catalog #: F-1297.0005BA
Supplier: Bachem Americas
CAS Number: 10433-52-0
Specifications
- Size:5 g
- Cat. no.:F-1297.0005BA
- Supplier No.:F-1297.0005
Specifications
About this item
Synonyms: H-D-Ser(Bzl)-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 195.218 g/mol
Formula:C₁₀H₁₃NO₃
MW:195.218 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00065936
CAS Number:10433-52-0
EINECS:233-916-6
Customers Who Bought This Also Bought