H-D-Asp(OtBu)-OH
Catalog #: F-1275.0005BA
Supplier: Bachem Americas
CAS Number: 64960-75-4
Specifications
- Size:5 g
- Cat. no.:F-1275.0005BA
- Supplier No.:F-1275.0005
Specifications
About this item
Synonyms: D-Aspartic acid 4-tert-butyl ester
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 189,211 g/mol
Formula:C₈H₁₅NO₄
Storage Temperature:Freezer
MDL Number:MFCD00077101
CAS Number:64960-75-4
Customers Who Bought This Also Bought