l-Isoasparagine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: H-Asp-NH₂
Formula:C₄H₈N₂O₃
MW:132.119 g/mol
MDL Number:MFCD00056118
CAS Number:28057-52-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 132.119 g/mol
Storage Temperature:Ambient
Customers Who Bought This Also Bought