(S)-Benzyl-2,6-diaminohexanoate bis(4-methylbenzenesulfonate)
Catalog #: E-3715.0001BA
Supplier: Bachem Americas
CAS Number: 16259-78-2
Specifications
- Size:1 g
- Cat. no.:E-3715.0001BA
- Supplier No.:E-3715.0001
Specifications
About this item
Synonyms: (S)-Benzyl 2,6-diaminohexanoate bis(4-methylbenzenesulfonate),(S)-Benzyl 2,6-diaminohexanoate bis(4-methylbenzenesulphonate)
:(S)-Benzyl-2,6-diaminohexanoate bis(4-methylbenzenesulfonate)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 580.711 g/mol
Formula:C₂₇H₃₆N₂O₈S₂
MW:580.711 g/mol
Storage Temperature:Freezer
MDL Number:MFCD08458627
CAS Number:16259-78-2
EINECS:240-366-0
Customers Who Bought This Also Bought