L-Asparagine methyl ester hydrochloride
Catalog #: E-3485.0001BA
Supplier: Bachem Americas
CAS Number: 57461-34-4
Specifications
- Size:1 g
- Cat. no.:E-3485.0001BA
- Supplier No.:E-3485.0001
Specifications
About this item
Synonyms: (S)-Methyl 2,4-diamino-4-oxobutanoate hydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 182.604 g/mol
:L-Asparagine methyl ester hydrochloride
Formula:C₅H₁₁ClN₂O₃
MW:182.604 g/mol
Storage Temperature:Freezer
MDL Number:MFCD01317794
CAS Number:57461-34-4
EINECS:260-748-0
Customers Who Bought This Also Bought