L-Threonine-tert-butyl ester hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (2S,3R)-tert-Butyl 2-amino-3-hydroxybutanoate hydrochloride
Formula:C₈H₁₇NO₃
MW:211.69 g/mol
MDL Number:MFCD00672361
CAS Number:69320-90-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 211.69 g/mol
:L-Threonine-tert-butyl ester hydrochloride
Storage Temperature:Freezer
Customers Who Bought This Also Bought