AC-ARG(PMC)-OH
Supplier: Bachem Americas
Certificates
About this item
Formula:C₂₂H₃₄N₄O₆S
MDL Number:MFCD00236744
CAS Number:163277-77-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 482,596 g/mol
:AC-ARG(PMC)-OH
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought