Glycine
Catalog #: E-3000.0500BA
Supplier: Bachem Americas
CAS Number: 56-40-6
Specifications
- Size:500 g
- Cat. no.:E-3000.0500BA
- Supplier No.:E-3000.0500
Specifications
About this item
Synonyms: Aminoacetic acid,Glycocoll
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 83 °C
:Flash Pt:
:Flash Pt:
:M.W. 75.07 g/mol
:Glycine
EINECS:200-272-2
:83
:181
Formula:NH₂CH₂COOH
MW:75.07 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00008131
CAS Number:56-40-6
Customers Who Bought This Also Bought