(S)-Methyl-2-amino-3-(1-trityl-1H-imidazol-4-yl)propanoate hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: H-His(Trt)-OMe.HCl,H-His(Trt)-OMe·HCl
Formula:C₂₆H₂₅N₃O₂·HCl
MW:447.963 g/mol
MDL Number:MFCD00237100
CAS Number:32946-56-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 447.963 g/mol
:(S)-Methyl-2-amino-3-(1-trityl-1H-imidazol-4-yl)propanoate hydrochloride
Storage Temperature:Freezer
Customers Who Bought This Also Bought