1-Methyl-L-aspartate
Supplier: Bachem Americas
Certificates
About this item
Synonyms: L-Aspartic Acid 1-Methyl Ester,H-Asp-OMe
Formula:C₅H₉NO₄
MW:147.13 g/mol
MDL Number:MFCD00080289
CAS Number:17812-32-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 147.13 g/mol
:1-Methyl-L-aspartate
Storage Temperature:Freezer
Customers Who Bought This Also Bought