N-Me-Phe-Obzl p-Tosylate
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-Me-Phe-OBzl·p-tosylate
Formula:C₂₄H₂₇NO₅S
MW:441.542 g/mol
MDL Number:MFCD00238321
CAS Number:40298-25-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 441.542 g/mol
Storage Temperature:Ambient
Customers Who Bought This Also Bought