(S)-2-(Methylamino)succinic acid
Catalog #: E-2745.0250BA
Supplier: Bachem Americas
CAS Number: 4226-18-0
Specifications
- Size:250 mg
- Cat. no.:E-2745.0250BA
- Supplier No.:E-2745.0250
Specifications
About this item
Synonyms: N-Me-Asp-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 147.13 g/mol
Formula:C₅H₉NO₄
MW:147.13 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00069561
CAS Number:4226-18-0
Customers Who Bought This Also Bought