Nε-acetyl-L-lysine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: L-Lys(Ac)-OH,H-Lys(Ac)-OH
Formula:C₈H₁₆N₂O₃
MW:188.23 g/mol
MDL Number:MFCD00002639
CAS Number:692-04-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 188.23 g/mol
Storage Temperature:Ambient
EINECS:211-725-9
Customers Who Bought This Also Bought