L-Glutamic acid diethyl ester hydrochloride
Catalog #: E-2670.0025BA
Supplier: Bachem Americas
CAS Number: 1118-89-4
Specifications
- Size:25 g
- Cat. no.:E-2670.0025BA
- Supplier No.:E-2670.0025
Specifications
About this item
Synonyms: Diethyl L-glutamate,H-Glu(OEt)-OEt.HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 239.696 g/mol
Formula:C₉H₁₈ClNO₄
MW:239.696 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00012509
CAS Number:1118-89-4
EINECS:214-270-4
Customers Who Bought This Also Bought