(S)-Methyl-6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (S)-Methyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride
Formula:C₂₂H₂₇ClN₂O₄
MW:418.918 g/mol
MDL Number:MFCD00153447
CAS Number:201009-98-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 418.918 g/mol
:(S)-Methyl-6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride
Storage Temperature:Freezer
Customers Who Bought This Also Bought