H-Val-OBzl·HCl
Catalog #: E-2575.0025BA
Supplier: Bachem Americas
CAS Number: 2462-34-2
Specifications
- Size:25 g
- Cat. no.:E-2575.0025BA
- Supplier No.:E-2575.0025
Specifications
About this item
Synonyms: L-Valine Benzyl Ester Hydrochloride,(S)-benzyl 2-amino-3-methylbutanoate hydrochloride,benzyl (2S)-2-amino-3-methylbutanoate hydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 243.731 g/mol
:H-Val-OBzl·HCl
MW:243.731 g/mol
Storage Temperature:Refrigerator
CAS Number:2462-34-2
:MFCD22422648
Formula:C₁₂H₁₈ClNO₂
MDL Number:MFCD00058010
Customers Who Bought This Also Bought