L-Tyrosine methyl ester hydrochloride
Catalog #: E-2555.0100BA
Supplier: Bachem Americas
CAS Number: 3417-91-2
Specifications
- Size:100 g
- Cat. no.:E-2555.0100BA
- Supplier No.:E-2555.0100
Specifications
About this item
Synonyms: L-Methyltyrosinate hydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 231.68 g/mol
:L-Tyrosine methyl ester hydrochloride
Formula:C₁₀H₁₃NO₃
MW:231.68 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD00012607
CAS Number:3417-91-2
EINECS:222-313-3
Customers Who Bought This Also Bought