L-serine methyl ester hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: H-Ser-OMe.HCl
Formula:C₄H₉NO₃
MW:155.58 g/mol
MDL Number:MFCD00063680
CAS Number:5680-80-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 155.58 g/mol
:L-serine methyl ester hydrochloride
Storage Temperature:Refrigerator
EINECS:227-140-7
Customers Who Bought This Also Bought