tert-Butyl-2-(methylamino)acetate hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Sar-OtBu·HCl,tert-Butyl sarcosinate hydrochloride,Sarcosine tert-butyl ester hydrochloride
CAS Number:136088-69-2
:5616-81-9
Formula:C₇H₁₆ClNO₂
MW:181.660 g/mol
MDL Number:MFCD00080936
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 181.660 g/mol
:tert-Butyl-2-(methylamino)acetate hydrochloride
Storage Temperature:Freezer
Customers Who Bought This Also Bought