(S)-Methyl-2-amino-4-methylpentanoate hydrochloride
Catalog #: E-2080.0100BA
Supplier: Bachem Americas
CAS Number: 7517-19-3
Specifications
- Size:100 g
- Cat. no.:E-2080.0100BA
- Supplier No.:E-2080.0100
Specifications
About this item
Synonyms: H-Leu-OMe·HCl,H-Leu-OMe.HCl
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 181.7 g/mol
:(S)-Methyl-2-amino-4-methylpentanoate hydrochloride
Formula:C₇H₁₅NO₂·HCl
MW:181.7 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD00012494
CAS Number:7517-19-3
EINECS:231-375-0
Customers Who Bought This Also Bought