L(+)-Histidine
Catalog #: E-2010.0250BA
Supplier: Bachem Americas
CAS Number: 71-00-1
Specifications
- Size:250 g
- Cat. no.:E-2010.0250BA
- Supplier No.:E-2010.0250
Specifications
About this item
Synonyms: L-3-Imidazol-4-ylalanine,(S)-(+)-Histidine,(S)-2-Amino-3-(4-imidazolyl)propionic acid,H-His-OH,NSC 137773
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 272 to 273 °C
:Flash Pt:
:Flash Pt:
:M.W. 155.16 g/mol
:L(+)-Histidine
Formula:C₆H₉N₃O₂
MW:155.16 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00064315
CAS Number:71-00-1
EINECS:200-745-3
Customers Who Bought This Also Bought