Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Methyl glycinate hydrochloride
Formula:C₃H₈ClNO₂
MW:125.56 g/mol
MDL Number:MFCD00012870
CAS Number:5680-79-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 125.56 g/mol
Storage Temperature:Ambient
EINECS:227-139-1
Customers Who Bought This Also Bought