Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl)
Supplier: Bachem Americas
Certificates
About this item
Synonyms: H-Gly-OtBu.HCl
Formula:C₆H₁₄ClNO₂
MW:167.64 g/mol
MDL Number:MFCD00058255
CAS Number:27532-96-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 167.64 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought