2,5-Piperazinedione
Catalog #: E-1965.0025BA
Supplier: Bachem Americas
CAS Number: 106-57-0
Specifications
- Size:25 g
- Cat. no.:E-1965.0025BA
- Supplier No.:E-1965.0025
Specifications
About this item
Synonyms: Glycine anhydride,Piperazine-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 114.1 g/mol
Formula:C₄H₆N₂O₂
MW:114.1 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00006009
CAS Number:106-57-0
Customers Who Bought This Also Bought