L(+)-Glutamine
Catalog #: E-1915.0100BA
Supplier: Bachem Americas
CAS Number: 56-85-9
Specifications
- Size:100 g
- Cat. no.:E-1915.0100BA
- Supplier No.:E-1915.0100
Specifications
About this item
Synonyms: Gln,L-Glutamine,H-Gln-OH,(S)-(+)-Glutamine,(S)-2,5-Diamino-5-oxopentanoic acid,Levoglutamide
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 185 to 186 °C
:Flash Pt:
:Flash Pt:
:M.W. 146.15 g/mol
:L(+)-Glutamine
Formula:C₅H₁₀N₂O₃
MW:146.15 g/mol
MDL Number:MFCD00008044
EINECS:200-292-1
:355
Storage Temperature:Refrigerator
CAS Number:56-85-9
Customers Who Bought This Also Bought