Dimethyl-L-glutamate hydrochloride
Catalog #: E-1905.0050BA
Supplier: Bachem Americas
CAS Number: 23150-65-4
Specifications
- Size:50 g
- Cat. no.:E-1905.0050BA
- Supplier No.:E-1905.0050
Specifications
About this item
Synonyms: L-Glutamic Acid Dimethyl Ester Hydrochloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 211.64 g/mol
Formula:C₇H₁₃NO₄
MW:211.64 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD00038879
CAS Number:23150-65-4
EINECS:245-461-0
Customers Who Bought This Also Bought